Assay Details
| Gene Symbol: RANBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300865.1 | NP_001287794.1 | 1 - 2 | 251 | 107 | |
| NM_007320.2 | NP_015559.2 | 1 - 2 | 251 | 107 | ||
| GenBank mRNA | AK314343.1 | - | 1 - 2 | 82 | 107 | |
| BM786367.1 | - | 1 - 2 | 251 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK314343.1, NM_001300865.1, NM_007320.2 transcripts.
Set Membership: |
|

RANBP3
Genomic Map