Assay Details
| Gene Symbol: ERBB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005862.2 | NP_001005862.1 | 4 - 5 | 561 | 79 | |
| NM_001289938.1 | NP_001276867.1 | 4 - 5 | 561 | 79 | ||
| GenBank mRNA | BC110392.1 | - | 4 - 5 | 526 | 79 | |
| BI755753.1 | - | 4 - 5 | 526 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001005862.2, NM_001289938.1 transcripts.
Set Membership: |
|

ERBB2
Genomic Map