Assay Details
| Gene Symbol: CALB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001740.4 | NP_001731.2 | 3 - 4 | 347 | 133 | |
| NM_007088.3 | NP_009019.1 | 3 - 4 | 347 | 133 | ||
| NR_027910.2 | 3 - 4 | 347 | 133 | |||
| GenBank mRNA | AK222495.1 | - | 3 - 4 | 337 | 133 | |
| AK222648.1 | - | 3 - 4 | 337 | 133 | ||
| BC015484.2 | - | 3 - 4 | 322 | 3847342 | 133 | |
| DA060805.1 | - | 3 - 4 | 347 | 133 | ||
| X56667.1 | - | 3 - 4 | 310 | 133 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000589.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16008549.hg.1 | ENST00000302628.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008549.hg.1 | ENST00000349553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008549.hg.1 | ENST00000490520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008549.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008549.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008549.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008549.hg.1 | TC16000589.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000467817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000467817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000467817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000467817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000490520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000490520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000490520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000490520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000490520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000490520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000490520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000490520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600162555.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162555.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162555.hg.1 | ENST00000490520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162555.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162555.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600162555.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600162555.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600162555.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162555.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162555.hg.1 | ENST00000490520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162555.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162555.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600162555.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600162555.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600162555.hg.1 | TC1600008301.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000302628.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000349553.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000467817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000467817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000467817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000467817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000490520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000490520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000490520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000490520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000490520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000490520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000490520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000490520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | ENST00000562305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_001740.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NM_007088.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NR_027910.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008301.hg.1 | NR_027910.2 | Refseq |

CALB2
Genomic Map