Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009727.hg.1 |
ENST00000375815.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009727.hg.1 |
ENST00000375820.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009727.hg.1 |
ENST00000397198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009727.hg.1 |
NM_001845.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009727.hg.1 |
TC13000872.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300165185.hg.1 |
ENST00000375820.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300165185.hg.1 |
NM_001845.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165185.hg.1 |
ENST00000375820.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300165185.hg.1 |
NM_001845.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165185.hg.1 |
TC1300009781.hg.1 |
Clariom D TC |