Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1300165213.hg.1 |
ENST00000375820.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300165213.hg.1 |
ENST00000543140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300165213.hg.1 |
ENST00000615732.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300165213.hg.1 |
NM_001303110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165213.hg.1 |
NM_001845.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165213.hg.1 |
ENST00000375820.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300165213.hg.1 |
ENST00000543140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300165213.hg.1 |
ENST00000615732.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300165213.hg.1 |
NM_001303110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165213.hg.1 |
NM_001845.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165213.hg.1 |
TC1300009781.hg.1 |
Clariom D TC |