Assay Details
| Gene Symbol: KIF5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004984.2 | NP_004975.2 | 13 - 14 | 1574 | 75 | |
| GenBank mRNA | AB210045.1 | - | 13 - 14 | 1399 | 75 | |
| AB290174.1 | - | 13 - 14 | 1640 | 75 | ||
| AK294881.1 | - | 10 - 11 | 1327 | 75 | ||
| AK315202.1 | - | 13 - 14 | 1594 | 75 | ||
| BC146670.1 | - | 13 - 14 | 1577 | 40148192 | 75 | |
| BC150208.1 | - | 13 - 14 | 1577 | 40148890 | 75 | |
| U06698.1 | - | 13 - 14 | 1514 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000534.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
retrograde vesicle-mediated transport, Golgi to ER
microtubule-based movement
chemical synaptic transmission
axon guidance
protein localization
antigen processing and presentation of exogenous peptide antigen via MHC class II
cytoskeleton-dependent intracellular transport
motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
retrograde vesicle-mediated transport, Golgi to ER
microtubule-based movement
chemical synaptic transmission
axon guidance
protein localization
antigen processing and presentation of exogenous peptide antigen via MHC class II
cytoskeleton-dependent intracellular transport
motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12007086.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007086.hg.1 | ENST00000455537.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007086.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007086.hg.1 | TC12000534.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000552227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000552227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000552227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000552227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000552227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000552227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000552227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000552227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200734.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200734.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200734.hg.1 | ENST00000286452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200734.hg.1 | NM_004984.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200734.hg.1 | TC1200012651.hg.1 | Clariom D TC |

KIF5A
Genomic Map