Assay Details
Gene Symbol: KIF5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004984.2 | NP_004975.2 | 17 - 18 | 2229 | 71 | |
GenBank mRNA | AB210045.1 | - | 17 - 18 | 2054 | 71 | |
AB290174.1 | - | 17 - 18 | 2295 | 71 | ||
AK294881.1 | - | 14 - 15 | 1982 | 71 | ||
BC146670.1 | - | 17 - 18 | 2232 | 40148192 | 71 | |
BC150208.1 | - | 17 - 18 | 2232 | 40148890 | 71 | |
U06698.1 | - | 17 - 18 | 2169 | 71 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000534.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
retrograde vesicle-mediated transport, Golgi to ER
microtubule-based movement
chemical synaptic transmission
axon guidance
protein localization
antigen processing and presentation of exogenous peptide antigen via MHC class II
cytoskeleton-dependent intracellular transport
motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
retrograde vesicle-mediated transport, Golgi to ER
microtubule-based movement
chemical synaptic transmission
axon guidance
protein localization
antigen processing and presentation of exogenous peptide antigen via MHC class II
cytoskeleton-dependent intracellular transport
motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12007089.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007089.hg.1 | ENST00000455537.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007089.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007089.hg.1 | TC12000534.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12007090.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007090.hg.1 | ENST00000455537.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007090.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007090.hg.1 | TC12000534.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12007091.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007091.hg.1 | ENST00000455537.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007091.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007091.hg.1 | TC12000534.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000552227.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000552227.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000552227.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | ENST00000552227.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000552227.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000552227.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000552227.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | ENST00000552227.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012651.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200737.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200737.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200737.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200737.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200737.hg.1 | TC1200012651.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200200739.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200739.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200739.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200739.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200739.hg.1 | TC1200012651.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200200738.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200738.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200738.hg.1 | ENST00000286452.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200738.hg.1 | NM_004984.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200738.hg.1 | TC1200012651.hg.1 | Clariom D TC |
