Assay Details
| Gene Symbol: ALDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000690.3 | NP_000681.2 | 9 - 10 | 1180 | 72 | |
| NM_001204889.1 | NP_001191818.1 | 8 - 9 | 1039 | 72 | ||
| GenBank mRNA | AK223373.1 | - | 9 - 10 | 1122 | 72 | |
| AK301375.1 | - | 8 - 9 | 977 | 72 | ||
| AK314856.1 | - | 9 - 10 | 1101 | 72 | ||
| AY621070.1 | - | 9 - 10 | 1145 | 72 | ||
| BC002967.1 | - | 9 - 10 | 1130 | 3543343 | 72 | |
| BC071839.1 | - | 9 - 10 | 1089 | 5477768 | 72 | |
| K03001.1 | - | 6 - 7 | 730 | 72 | ||
| M26760.1 | - | 6 - 7 | 730 | 72 | ||
| M54931.1 | - | 4 - 5 | 445 | 72 | ||
| X05409.1 | - | 9 - 10 | 1117 | 72 | ||
| Y00109.1 | - | 9 - 10 | 1108 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12003212.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000551450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000551450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011485.hg.1 | TC12003212.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011485.hg.1 | ENST00000261733.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011485.hg.1 | ENST00000416293.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011485.hg.1 | ENST00000552234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011485.hg.1 | ENST00000553044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011485.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011485.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201651.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201651.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201651.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201651.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201651.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201651.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201651.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201651.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201651.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201651.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201651.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201651.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201651.hg.1 | TC1200012707.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000551450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000551450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |

ALDH2
Genomic Map