Assay Details
| Gene Symbol: CABP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145200.3 | NP_660201.1 | 2 - 3 | 472 | 73 | |
| XM_005274114.3 | XP_005274171.2 | 3 - 4 | 477 | 73 | ||
| XM_011545181.2 | XP_011543483.1 | 3 - 4 | 477 | 73 | ||
| GenBank mRNA | AY039217.1 | - | 2 - 3 | 472 | 73 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000692.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11008996.hg.1 | ENST00000325656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008996.hg.1 | ENST00000438189.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008996.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008996.hg.1 | TC11000692.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000602912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000602912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100159954.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159954.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159954.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159954.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100159954.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100159954.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100159954.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159954.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159954.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159954.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100159954.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100159954.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100159954.hg.1 | TC1100008138.hg.1 | Clariom D TC |

CABP4
Genomic Map