Assay Details
| Gene Symbol: PICALM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206946.1 | NP_001193875.1 | 12 - 13 | 1582 | 108 | |
| XM_006718699.2 | XP_006718762.1 | 12 - 13 | 1566 | 108 | ||
| XM_006718700.2 | XP_006718763.1 | 12 - 13 | 1566 | 108 | ||
| XM_006718701.3 | XP_006718764.1 | 12 - 13 | 1566 | 108 | ||
| XM_017018381.1 | XP_016873870.1 | 12 - 13 | 1566 | 108 | ||
| GenBank mRNA | AB210017.1 | - | 12 - 13 | 1582 | 108 | |
More Information
Set Membership: |
|

PICALM
Genomic Map