Assay Details
| Gene Symbol: GPCPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005260758.4 | XP_005260815.1 | 2 - 3 | 332 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005260758.4 transcripts.
Set Membership: |
|

GPCPD1
Genomic Map