Assay Details
Gene Symbol: B4GALT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017026088.1 | XP_016881577.1 | 1 - 2 | 1377 | 105 | |
XM_017026091.1 | XP_016881580.1 | 1 - 2 | 119 | 105 | ||
GenBank mRNA | AF069054.1 | - | 1 - 2 | 257 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017026091.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000579372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000579372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | ENST00000579372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000237019.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000306851.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000383131.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000578114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000579372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000579372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | ENST00000579372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008399.hg.1 | NM_004775.3 | Refseq |
