Assay Details
Gene Symbol: CLSTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022131.2 | NP_071414.2 | 14 - 15 | 2546 | 101 | |
XM_017007022.1 | XP_016862511.1 | 14 - 15 | 2587 | 101 | ||
GenBank mRNA | AJ278018.1 | - | 15 - 16 | 2366 | 101 | |
AK056411.1 | - | 8 - 9 | 1184 | 101 | ||
AK315308.1 | - | 15 - 16 | 2546 | 101 | ||
AY753303.1 | - | 15 - 16 | 2356 | 101 | ||
BC004871.2 | - | 2 - 3 | 204 | 101 | ||
BC103496.1 | - | 15 - 16 | 2366 | 40008317 | 101 | |
BC103506.1 | - | 14 - 15 | 2365 | 40008319 | 101 | |
BC104485.1 | - | 15 - 16 | 2363 | 40008318 | 101 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000754.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03013708.hg.1 | TC03000754.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03013708.hg.1 | ENST00000458420.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03013708.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300164319.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300164319.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300164319.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300164319.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300164319.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300164319.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300164319.hg.1 | TC0300008979.hg.1 | Clariom D TC |
