Assay Details
Gene Symbol: CLSTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022131.2 | NP_071414.2 | 5 - 6 | 983 | 67 | |
XM_017007022.1 | XP_016862511.1 | 5 - 6 | 1024 | 67 | ||
GenBank mRNA | AJ278018.1 | - | 5 - 6 | 803 | 67 | |
AK096432.1 | - | 5 - 6 | 983 | 67 | ||
AK315308.1 | - | 5 - 6 | 983 | 67 | ||
AY753303.1 | - | 5 - 6 | 793 | 67 | ||
BC103496.1 | - | 5 - 6 | 803 | 40008317 | 67 | |
BC103506.1 | - | 5 - 6 | 802 | 40008319 | 67 | |
BC104485.1 | - | 5 - 6 | 800 | 40008318 | 67 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000754.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03013698.hg.1 | TC03000754.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03013698.hg.1 | ENST00000458420.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03013698.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300164309.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300164309.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300164309.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300164309.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300164309.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300164309.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300164309.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300164309.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300164309.hg.1 | TC0300008979.hg.1 | Clariom D TC |
