Assay Details
Gene Symbol: CLSTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022131.2 | NP_071414.2 | 6 - 7 | 1164 | 106 | |
XM_017007022.1 | XP_016862511.1 | 6 - 7 | 1205 | 106 | ||
GenBank mRNA | AJ278018.1 | - | 6 - 8 | 106 | ||
AK096432.1 | - | 6 - 8 | 106 | |||
AK315308.1 | - | 6 - 8 | 106 | |||
AY753303.1 | - | 6 - 8 | 106 | |||
BC103496.1 | - | 6 - 8 | 40008317 | 106 | ||
BC103506.1 | - | 6 - 7 | 983 | 40008319 | 106 | |
BC104485.1 | - | 6 - 8 | 40008318 | 106 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000754.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03013699.hg.1 | TC03000754.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03013699.hg.1 | ENST00000458420.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03013699.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300164310.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300164310.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300164310.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300164310.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300164310.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300164310.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300164310.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300164310.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300164310.hg.1 | TC0300008979.hg.1 | Clariom D TC |
