Assay Details
Gene Symbol: CUZD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF305835.1 | - | 1 - 2 | 700 | 154 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF305835.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
