Assay Details
Gene Symbol: SHTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127211.2 | NP_001120683.1 | 13 - 14 | 1807 | 99 | |
NM_001258298.1 | NP_001245227.1 | 12 - 13 | 1754 | 99 | ||
NM_001258299.1 | NP_001245228.1 | 13 - 14 | 1807 | 99 | ||
NM_001258300.1 | NP_001245229.1 | 14 - 15 | 1597 | 99 | ||
XM_005270002.3 | XP_005270059.1 | 13 - 14 | 1625 | 99 | ||
XM_006717931.3 | XP_006717994.1 | 13 - 14 | 1625 | 99 | ||
XM_017016462.1 | XP_016871951.1 | 13 - 14 | 1317 | 99 | ||
GenBank mRNA | AK091381.1 | - | 3 - 4 | 440 | 99 | |
AK091578.1 | - | 12 - 13 | 1513 | 99 | ||
AK095419.1 | - | 3 - 4 | 266 | 99 | ||
AK302705.1 | - | 14 - 15 | 1597 | 99 | ||
AK304865.1 | - | 12 - 13 | 1401 | 99 | ||
BC022348.1 | - | 2 - 3 | 149 | 99 | ||
BC032303.1 | - | 13 - 14 | 1365 | 5169981 | 99 | |
CR749417.1 | - | 13 - 14 | 1415 | 99 | ||
CX783645.1 | - | 4 - 5 | 378 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC022348.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
