Assay Details
| Gene Symbol: WBP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083913.1 | NP_001077382.1 | 1 - 2 | 175 | 99 | |
| XM_017016360.1 | XP_016871849.1 | 1 - 2 | 1437 | 99 | ||
| GenBank mRNA | AK300871.1 | - | 1 - 2 | 220 | 99 | |
| AK308843.1 | - | 1 - 2 | 184 | 99 | ||
| AK316452.1 | - | 1 - 2 | 187 | 99 | ||
| CX163170.1 | - | 1 - 2 | 121 | 99 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000764.hg
Additional Information:
This assay is in the 5'UTR region of the XM_017016360.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500010679.hg.1 | ENST00000560907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013440.hg.1 | ENST00000515260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009095.hg.1 | ENST00000369889.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009095.hg.1 | ENST00000448841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009095.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009095.hg.1 | NM_017787.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009095.hg.1 | TC10000764.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006437.hg.1 | ENST00000586141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006450.hg.1 | ENST00000437905.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009091.hg.1 | ENST00000448841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009091.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009091.hg.1 | TC10000764.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013133.hg.1 | ENST00000505043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006436.hg.1 | ENST00000523033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | ENST00000369889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | ENST00000369889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | ENST00000369889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | ENST00000369889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | ENST00000369889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | ENST00000369889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | ENST00000369889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | NM_017787.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | NM_017787.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | NM_017787.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | NM_017787.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | NM_017787.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | NM_017787.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | NM_017787.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008750.hg.1 | NM_017787.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014047.hg.1 | ENST00000617401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006433.hg.1 | ENST00000604449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006432.hg.1 | ENST00000519787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006460.hg.1 | ENST00000438434.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013133.hg.1 | ENST00000505043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006432.hg.1 | ENST00000519787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159904.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159904.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159904.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159904.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159904.hg.1 | TC1000008750.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800006436.hg.1 | ENST00000523033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010679.hg.1 | ENST00000560907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006460.hg.1 | ENST00000438434.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014047.hg.1 | ENST00000617401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013440.hg.1 | ENST00000515260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159905.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159905.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159905.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159905.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159905.hg.1 | TC1000008750.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1900006437.hg.1 | ENST00000586141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | ENST00000369889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | ENST00000369889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | ENST00000369889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | ENST00000369889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | ENST00000369889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | ENST00000369889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | ENST00000369889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | NM_017787.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | NM_017787.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | NM_017787.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | NM_017787.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | NM_017787.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | NM_017787.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | NM_017787.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008750.hg.1 | NM_017787.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159910.hg.1 | ENST00000369889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159910.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159910.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159910.hg.1 | NM_017787.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159910.hg.1 | ENST00000369889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159910.hg.1 | ENST00000448841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159910.hg.1 | NM_001083913.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159910.hg.1 | NM_017787.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159910.hg.1 | TC1000008750.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600006433.hg.1 | ENST00000604449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006450.hg.1 | ENST00000437905.2 | Ensembl |

WBP1L
Genomic Map