Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005661.hg.1 |
ENST00000211998.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005661.hg.1 |
ENST00000372755.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005661.hg.1 |
ENST00000415462.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005661.hg.1 |
ENST00000417648.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005661.hg.1 |
ENST00000537043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005661.hg.1 |
NM_003373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005661.hg.1 |
NM_014000.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005661.hg.1 |
TC10000476.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000154753.hg.1 |
ENST00000211998.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154753.hg.1 |
ENST00000372755.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154753.hg.1 |
ENST00000436396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154753.hg.1 |
ENST00000623461.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154753.hg.1 |
ENST00000624354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154753.hg.1 |
NM_003373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000154753.hg.1 |
NM_014000.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000154753.hg.1 |
ENST00000211998.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154753.hg.1 |
ENST00000372755.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154753.hg.1 |
ENST00000436396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154753.hg.1 |
ENST00000623461.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154753.hg.1 |
ENST00000624354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154753.hg.1 |
NM_003373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000154753.hg.1 |
NM_014000.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000154753.hg.1 |
TC1000008056.hg.1 |
Clariom D TC |