Assay Details
| Gene Symbol: TCF7L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146284.1 | NP_001139756.1 | 6 - 7 | 1215 | 69 | |
| NM_001198527.1 | NP_001185456.1 | 6 - 7 | 1215 | 69 | ||
| XM_005270088.1 | XP_005270145.1 | 6 - 7 | 1246 | 69 | ||
| XM_005270102.1 | XP_005270159.1 | 7 - 8 | 1315 | 69 | ||
| XM_005270104.1 | XP_005270161.1 | 6 - 7 | 1246 | 69 | ||
| XM_011540110.1 | XP_011538412.1 | 7 - 8 | 1390 | 69 | ||
| XM_017016585.1 | XP_016872074.1 | 8 - 9 | 1459 | 69 | ||
| XM_017016587.1 | XP_016872076.1 | 8 - 9 | 1459 | 69 | ||
| XM_017016591.1 | XP_016872080.1 | 7 - 8 | 1315 | 69 | ||
| XM_017016596.1 | XP_016872085.1 | 7 - 8 | 1315 | 69 | ||
| GenBank mRNA | BC032656.1 | - | 6 - 7 | 804 | 5533185 | 69 |
| FJ010165.1 | - | 6 - 7 | 747 | 69 | ||
| FJ010168.1 | - | 6 - 7 | 805 | 69 | ||
| HM352840.1 | - | 7 - 8 | 777 | 69 | ||
| HM352843.1 | - | 6 - 7 | 708 | 69 | ||
More Information
Set Membership: |
|

TCF7L2
Genomic Map