Assay Details
| Gene Symbol: TCF7L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011540109.1 | XP_011538411.1 | 14 - 15 | 2065 | 64 | |
| XM_011540110.1 | XP_011538412.1 | 13 - 14 | 1969 | 64 | ||
| XM_011540111.1 | XP_011538413.1 | 12 - 13 | 1837 | 64 | ||
| XM_017016584.1 | XP_016872073.1 | 14 - 15 | 2062 | 64 | ||
| XM_017016585.1 | XP_016872074.1 | 14 - 15 | 2053 | 64 | ||
| XM_017016586.1 | XP_016872075.1 | 14 - 15 | 2050 | 64 | ||
| XM_017016587.1 | XP_016872076.1 | 14 - 15 | 2038 | 64 | ||
| XM_017016588.1 | XP_016872077.1 | 13 - 14 | 1996 | 64 | ||
| XM_017016590.1 | XP_016872079.1 | 13 - 14 | 1921 | 64 | ||
| XM_017016591.1 | XP_016872080.1 | 13 - 14 | 1909 | 64 | ||
| XM_017016592.1 | XP_016872081.1 | 13 - 14 | 1906 | 64 | ||
| XM_017016594.1 | XP_016872083.1 | 12 - 13 | 1852 | 64 | ||
| GenBank mRNA | FJ010164.1 | - | 11 - 12 | 1194 | 64 | |
| FJ010165.1 | - | 12 - 13 | 1326 | 64 | ||
More Information
Set Membership: |
|

TCF7L2
Genomic Map