Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044103.hg.1 |
TC01002832.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044103.hg.1 |
ENST00000370571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044103.hg.1 |
ENST00000436319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044103.hg.1 |
NM_152890.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210183.hg.1 |
ENST00000370571.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210183.hg.1 |
ENST00000426639.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210183.hg.1 |
NM_152890.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210183.hg.1 |
ENST00000370571.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210183.hg.1 |
ENST00000426639.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210183.hg.1 |
NM_152890.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210183.hg.1 |
TC0100014790.hg.1 |
Clariom D TC |