Assay Details
Gene Symbol: LINC01010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038216.1 | 1 - 2 | 167 | 87 | ||
NR_038217.1 | 1 - 2 | 167 | 87 | |||
GenBank mRNA | AK055166.1 | - | 1 - 2 | 170 | 87 | |
DA419376.1 | - | 1 - 2 | 170 | 87 | ||
DA420003.1 | - | 1 - 2 | 169 | 87 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001006.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06011854.hg.1 | TC06001006.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06011854.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06011854.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06011854.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06011854.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06011854.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06011851.hg.1 | TC06001006.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06011851.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06011851.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06011851.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000456749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000456749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | ENST00000456749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06011855.hg.1 | TC06001006.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06011855.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06011856.hg.1 | TC06001006.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06011856.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06011856.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165322.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165322.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165322.hg.1 | TC0600009521.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600165321.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600165321.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165321.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165321.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165321.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165321.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600165321.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165321.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165321.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165321.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165321.hg.1 | TC0600009521.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600165323.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600165323.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600165323.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165323.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600165323.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600165323.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165323.hg.1 | TC0600009521.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600165320.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600165320.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165320.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165320.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165320.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165320.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600165320.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165320.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165320.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165320.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165320.hg.1 | TC0600009521.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000440090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000456749.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000456749.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | ENST00000456749.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009521.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165315.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600165315.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165315.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165315.hg.1 | ENST00000431422.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600165315.hg.1 | NR_038216.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165315.hg.1 | NR_038217.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165315.hg.1 | TC0600009521.hg.1 | Clariom D TC |
