Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010011.hg.1 |
TC09000775.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010011.hg.1 |
ENST00000338351.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010011.hg.1 |
ENST00000355699.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010011.hg.1 |
ENST00000356589.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010011.hg.1 |
ENST00000371916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010011.hg.1 |
ENST00000371929.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010011.hg.1 |
NM_139025.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010011.hg.1 |
NM_139026.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010011.hg.1 |
NM_139027.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161827.hg.1 |
ENST00000355699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161827.hg.1 |
ENST00000356589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161827.hg.1 |
ENST00000371929.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161827.hg.1 |
ENST00000474918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161827.hg.1 |
NM_139025.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161827.hg.1 |
NM_139026.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161827.hg.1 |
NM_139027.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161827.hg.1 |
ENST00000355699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161827.hg.1 |
ENST00000356589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161827.hg.1 |
ENST00000371929.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161827.hg.1 |
ENST00000474918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161827.hg.1 |
NM_139025.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161827.hg.1 |
NM_139026.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161827.hg.1 |
NM_139027.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161827.hg.1 |
TC0900009077.hg.1 |
Clariom D TC |