Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000619.hg.1 |
TC07000040.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000619.hg.1 |
ENST00000389531.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000619.hg.1 |
ENST00000404826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000619.hg.1 |
ENST00000446104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000619.hg.1 |
NM_001079653.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000619.hg.1 |
NM_152744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700145245.hg.1 |
ENST00000389531.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145245.hg.1 |
ENST00000404826.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145245.hg.1 |
ENST00000476701.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145245.hg.1 |
ENST00000615806.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145245.hg.1 |
NM_001079653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700145245.hg.1 |
NM_152744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700145245.hg.1 |
ENST00000389531.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145245.hg.1 |
ENST00000404826.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145245.hg.1 |
ENST00000476701.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145245.hg.1 |
ENST00000615806.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145245.hg.1 |
NM_001079653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700145245.hg.1 |
NM_152744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700145245.hg.1 |
TC0700006583.hg.1 |
Clariom D TC |