Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016291.hg.1 |
TC06001400.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016291.hg.1 |
NR_002722.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016291.hg.1 |
NR_024553.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016292.hg.1 |
TC06001400.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016292.hg.1 |
NR_002722.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016292.hg.1 |
NR_024553.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016294.hg.1 |
TC06001400.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016294.hg.1 |
NR_024553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175625.hg.1 |
NR_024553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175625.hg.1 |
NR_024553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175625.hg.1 |
TC0600011194.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600175616.hg.1 |
NR_002722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175616.hg.1 |
NR_024553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175616.hg.1 |
NR_002722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175616.hg.1 |
NR_024553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175616.hg.1 |
TC0600011194.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600175612.hg.1 |
NR_002722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175612.hg.1 |
NR_024553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175612.hg.1 |
NR_002722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175612.hg.1 |
NR_024553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175612.hg.1 |
TC0600011194.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600175622.hg.1 |
NR_024553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175622.hg.1 |
NR_024553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175622.hg.1 |
TC0600011194.hg.1 |
Clariom D TC |