Assay Details
Gene Symbol: AARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001605.2 | NP_001596.2 | 13 - 14 | 1897 | 64 | |
GenBank mRNA | AK299098.1 | - | 13 - 14 | 1897 | 64 | |
BC011451.1 | - | 13 - 14 | 1829 | 3450606 | 64 | |
D32050.1 | - | 13 - 14 | 1897 | 64 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16001234.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
hair follicle development
tRNA modification
tRNA aminoacylation for protein translation
alanyl-tRNA aminoacylation
regulation of translational fidelity
protein folding
tRNA processing
cerebellar Purkinje cell layer development
endoplasmic reticulum unfolded protein response
response to amino acid
negative regulation of neuron apoptotic process
neuromuscular process controlling balance
tRNA binding
aminoacyl-tRNA editing activity
alanine-tRNA ligase activity
ATP binding
zinc ion binding
amino acid binding
hair follicle development
tRNA modification
tRNA aminoacylation for protein translation
alanyl-tRNA aminoacylation
regulation of translational fidelity
protein folding
tRNA processing
cerebellar Purkinje cell layer development
endoplasmic reticulum unfolded protein response
response to amino acid
negative regulation of neuron apoptotic process
neuromuscular process controlling balance
tRNA binding
aminoacyl-tRNA editing activity
alanine-tRNA ligase activity
ATP binding
zinc ion binding
amino acid binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR16017036.hg.1 | ENST00000261772.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16017036.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16017036.hg.1 | TC16001234.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000564359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000564359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000564359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000564359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000564359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000564359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000565361.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000565361.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000565361.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000565361.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000565361.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000565361.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000565361.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000566969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000566969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000566969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000566969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000566969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000567490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000567490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000567490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000569790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000569790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000569790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000569790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000569825.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000569825.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000569825.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000569825.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000569825.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000569825.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | ENST00000569825.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000564359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000564359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000564359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000564359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000564359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000564359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000565361.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000565361.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000565361.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000565361.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000565361.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000565361.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000565361.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000566969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000566969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000566969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000566969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000566969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000567490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000567490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000567490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000569790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000569790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000569790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000569790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000569825.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000569825.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000569825.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000569825.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000569825.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000569825.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | ENST00000569825.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010752.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183105.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183105.hg.1 | ENST00000564359.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183105.hg.1 | ENST00000565361.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183105.hg.1 | ENST00000567490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183105.hg.1 | ENST00000569790.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183105.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183105.hg.1 | ENST00000261772.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183105.hg.1 | ENST00000564359.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183105.hg.1 | ENST00000565361.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183105.hg.1 | ENST00000567490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183105.hg.1 | ENST00000569790.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183105.hg.1 | NM_001605.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183105.hg.1 | TC1600010752.hg.1 | Clariom D TC |
