Assay Details
Gene Symbol: KCNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011541396.2 | XP_011539698.1 | 3 | 2422 | 72 | |
XM_011541397.2 | XP_011539699.1 | 4 | 2703 | 72 | ||
XM_011541398.2 | XP_011539700.1 | 3 | 2470 | 72 | ||
XM_011541399.2 | XP_011539701.1 | 3 | 2511 | 72 | ||
XM_011541400.2 | XP_011539702.1 | 3 | 2441 | 72 | ||
XM_017001213.1 | XP_016856702.1 | 3 | 2403 | 72 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
potassium ion transport
regulation of dopamine secretion
neuronal action potential
sensory perception of pain
optic nerve structural organization
regulation of ion transmembrane transport
regulation of circadian sleep/wake cycle, non-REM sleep
protein homooligomerization
potassium ion transmembrane transport
voltage-gated potassium channel activity
delayed rectifier potassium channel activity
potassium channel activity
protein binding
potassium ion transport
regulation of dopamine secretion
neuronal action potential
sensory perception of pain
optic nerve structural organization
regulation of ion transmembrane transport
regulation of circadian sleep/wake cycle, non-REM sleep
protein homooligomerization
potassium ion transmembrane transport
voltage-gated potassium channel activity
delayed rectifier potassium channel activity
potassium channel activity
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100009390.hg.1 | ENST00000578271.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000525120.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000525120.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000525120.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_004974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_004974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_004974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_004974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_004974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_004974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_004974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_004974.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015244.hg.1 | NM_004974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009390.hg.1 | ENST00000578271.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000316361.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000369770.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000485317.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000525120.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000525120.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000525120.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | ENST00000633222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_001204269.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_004974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_004974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_004974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_004974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_004974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_004974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_004974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_004974.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015244.hg.1 | NM_004974.3 | Refseq |
