| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014063.hg.1 |
ENST00000419476.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027762.hg.1 |
TC01001775.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027762.hg.1 |
ENST00000366988.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027762.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027762.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000472389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000472389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010145.hg.1 |
ENST00000503735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000472389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000472389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014063.hg.1 |
ENST00000419476.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010145.hg.1 |
ENST00000503735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184894.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184894.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184894.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184894.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184894.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184894.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184894.hg.1 |
TC0100011527.hg.1 |
Clariom D TC |