Assay Details
Gene Symbol: RIPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006871.3 | NP_006862.2 | 3 - 4 | 683 | 89 | |
GenBank mRNA | AF156884.1 | - | 3 - 4 | 473 | 89 | |
AK075275.1 | - | 3 - 4 | 637 | 89 | ||
AK296140.1 | - | 3 - 4 | 636 | 89 | ||
AY453693.1 | - | 3 - 4 | 473 | 89 | ||
AY494982.1 | - | 3 - 4 | 473 | 89 | ||
AY494983.1 | - | 3 - 4 | 473 | 89 | ||
BC062584.1 | - | 3 - 4 | 614 | 5213027 | 89 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14002317.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of T cell mediated cytotoxicity
regulation of adaptive immune response
cellular protein modification process
signal transduction
I-kappaB kinase/NF-kappaB signaling
positive regulation of phosphatase activity
activation of protein kinase activity
regulation of interferon-gamma production
T cell differentiation in thymus
NIK/NF-kappaB signaling
T cell homeostasis
regulation of activated T cell proliferation
protein autophosphorylation
lymph node development
spleen development
thymus development
positive regulation of NF-kappaB transcription factor activity
protein homooligomerization
protein heterooligomerization
positive regulation of ligase activity
positive regulation of oxidoreductase activity
positive regulation of necroptotic process
regulation of activation-induced cell death of T cells
necroptotic process
positive regulation of protein deacetylation
apoptotic signaling pathway
amyloid fibril formation
positive regulation of reactive oxygen species metabolic process
regulation of CD8-positive, alpha-beta cytotoxic T cell extravasation
positive regulation of intrinsic apoptotic signaling pathway
transcription coactivator activity
protein kinase activity
protein serine/threonine kinase activity
NF-kappaB-inducing kinase activity
protein binding
ATP binding
protein complex binding
identical protein binding
regulation of T cell mediated cytotoxicity
regulation of adaptive immune response
cellular protein modification process
signal transduction
I-kappaB kinase/NF-kappaB signaling
positive regulation of phosphatase activity
activation of protein kinase activity
regulation of interferon-gamma production
T cell differentiation in thymus
NIK/NF-kappaB signaling
T cell homeostasis
regulation of activated T cell proliferation
protein autophosphorylation
lymph node development
spleen development
thymus development
positive regulation of NF-kappaB transcription factor activity
protein homooligomerization
protein heterooligomerization
positive regulation of ligase activity
positive regulation of oxidoreductase activity
positive regulation of necroptotic process
regulation of activation-induced cell death of T cells
necroptotic process
positive regulation of protein deacetylation
apoptotic signaling pathway
amyloid fibril formation
positive regulation of reactive oxygen species metabolic process
regulation of CD8-positive, alpha-beta cytotoxic T cell extravasation
positive regulation of intrinsic apoptotic signaling pathway
transcription coactivator activity
protein kinase activity
protein serine/threonine kinase activity
NF-kappaB-inducing kinase activity
protein binding
ATP binding
protein complex binding
identical protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR14011191.hg.1 | TC14002317.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR14011191.hg.1 | ENST00000216274.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14011191.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14011193.hg.1 | TC14002317.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR14011193.hg.1 | ENST00000216274.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14011193.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554569.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554569.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554569.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557662.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557662.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557662.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557662.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400181987.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400181987.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400181987.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400181987.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400181987.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400181987.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400181987.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400181987.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400181987.hg.1 | TC1400010727.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400181989.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400181989.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400181989.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400181989.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400181989.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400181989.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400181989.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400181989.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400181989.hg.1 | TC1400010727.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554569.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554569.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554569.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557662.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557662.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557662.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557662.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
