Assay Details
Gene Symbol: CUZD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022034.5 | NP_071317.2 | 3 - 4 | 784 | 103 | |
NR_037912.1 | 2 - 3 | 633 | 103 | |||
GenBank mRNA | AK289445.1 | - | 2 - 3 | 315 | 103 | |
AY260047.1 | - | 3 - 4 | 784 | 103 | ||
AY260048.1 | - | 2 - 3 | 633 | 103 | ||
AY260049.1 | - | 2 - 3 | 633 | 103 | ||
BC136755.1 | - | 3 - 4 | 622 | 9020745 | 103 | |
BC144398.1 | - | 3 - 4 | 622 | 103 | ||
BX648572.1 | - | 3 - 4 | 467 | 103 |
Gene Symbol: FAM24B-CUZD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037915.1 | 5 - 6 | 1128 | 103 | ||
GenBank mRNA | AK300849.1 | - | 5 - 6 | 1117 | 103 | |
AY358556.1 | - | 5 - 6 | 1415 | 103 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10002953.hg
Additional Information:
This assay is in the 5'UTR region of the AK289445.1,AY260048.1,AY260049.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10022749.hg.1 | TC10002953.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10022749.hg.1 | ENST00000368899.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022749.hg.1 | ENST00000368900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022749.hg.1 | ENST00000368901.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022749.hg.1 | ENST00000368904.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022749.hg.1 | ENST00000392790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10022749.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10022749.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000338948.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012599.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000188051.hg.1 | NR_037915.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000188051.hg.1 | NR_037915.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000188051.hg.1 | TC1000012598.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000188068.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000188068.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000188068.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000188068.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000188068.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000188068.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000188068.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000188068.hg.1 | ENST00000368899.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000188068.hg.1 | ENST00000368900.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000188068.hg.1 | ENST00000368901.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000188068.hg.1 | ENST00000392790.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000188068.hg.1 | ENST00000545804.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000188068.hg.1 | NM_022034.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000188068.hg.1 | NR_037912.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000188068.hg.1 | TC1000012599.hg.1 | Clariom D TC |
