Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057456.hg.1 |
TC01003745.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057456.hg.1 |
ENST00000367157.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057456.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017064.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229242.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229242.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229242.hg.1 |
ENST00000367157.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229242.hg.1 |
NM_030952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229242.hg.1 |
TC0100017064.hg.1 |
Clariom D TC |