Assay Details
Gene Symbol: LRRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024512.4 | NP_078788.2 | 1 - 2 | 381 | 107 | |
XM_017007177.1 | XP_016862666.1 | 1 - 2 | 381 | 107 | ||
GenBank mRNA | AJ308569.2 | - | 1 - 2 | 155 | 107 | |
AK057808.1 | - | 1 - 2 | 223 | 107 | ||
AK223440.1 | - | 1 - 2 | 223 | 107 | ||
AK314658.1 | - | 1 - 2 | 144 | 107 | ||
AK315636.1 | - | 1 - 2 | 164 | 107 | ||
DA144445.1 | - | 1 - 2 | 381 | 107 | ||
DA342323.1 | - | 1 - 2 | 352 | 107 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001358.hg.1
Additional Information:
This assay is in the 5'UTR region of the AJ308569.2,AK057808.1,AK314658.1,AK315636.1,NM_024512.4,XM_017007177.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200014984.hg.1 | ENST00000430717.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014984.hg.1 | ENST00000430717.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03023203.hg.1 | TC03001358.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03023203.hg.1 | ENST00000395905.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03023203.hg.1 | ENST00000496388.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03023203.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03023201.hg.1 | TC03001358.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03023201.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03023201.hg.1 | ENST00000395905.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03023201.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03023204.hg.1 | TC03001358.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03023204.hg.1 | ENST00000395905.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03023204.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03023202.hg.1 | TC03001358.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03023202.hg.1 | ENST00000395905.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03023202.hg.1 | ENST00000496388.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03023202.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000469912.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000469912.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000469912.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000496388.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000496388.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | ENST00000496388.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014984.hg.1 | ENST00000430717.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014984.hg.1 | ENST00000430717.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000469912.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000469912.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000469912.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000496388.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000496388.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | ENST00000496388.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010952.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300179838.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179838.hg.1 | ENST00000469912.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179838.hg.1 | ENST00000496388.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179838.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300179838.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179838.hg.1 | ENST00000469912.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179838.hg.1 | ENST00000496388.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179838.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300179838.hg.1 | TC0300010952.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300179839.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179839.hg.1 | ENST00000496388.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179839.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300179839.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179839.hg.1 | ENST00000496388.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179839.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300179839.hg.1 | TC0300010952.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300179840.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179840.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300179840.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179840.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300179840.hg.1 | TC0300010952.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300179836.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179836.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179836.hg.1 | ENST00000469912.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179836.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300179836.hg.1 | ENST00000296144.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179836.hg.1 | ENST00000395905.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179836.hg.1 | ENST00000469912.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300179836.hg.1 | NM_024512.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300179836.hg.1 | TC0300010952.hg.1 | Clariom D TC |
