Assay Details
| Gene Symbol: RPL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000988.3 | NP_000979.1 | 2 - 3 | 122 | 88 | |
| GenBank mRNA | AK311792.1 | - | 2 - 3 | 112 | 88 | |
| AV712491.1 | - | 2 - 3 | 121 | 88 | ||
| BC001700.2 | - | 2 - 3 | 145 | 3050183 | 88 | |
| BC002588.2 | - | 2 - 3 | 102 | 3161438 | 88 | |
| BC007273.1 | - | 2 - 3 | 145 | 3161124 | 88 | |
| BC010026.2 | - | 2 - 3 | 149 | 3354227 | 88 | |
| BC098560.1 | - | 117 | 30415908 | 88 | ||
| L05094.1 | - | 1 - 2 | 102 | 88 | ||
| L19527.1 | - | 1 - 2 | 94 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000544.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300007618.hg.1 | ENST00000412199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007781.hg.1 | ENST00000411843.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010557.hg.1 | ENST00000434035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010536.hg.1 | ENST00000513087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007069.hg.1 | ENST00000477507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006624.hg.1 | ENST00000253788.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006624.hg.1 | NM_000988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006624.hg.1 | TC17000544.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000253788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000253788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000253788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000253788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000586277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000586277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000586277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000586277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000587478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000587478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000587478.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000588830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000588830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000588830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000589037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000589037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000589037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000589037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000589037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000589913.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000589913.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000589913.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000589913.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000589913.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000590864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000590864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000590864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000590864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | NM_000988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | NM_000988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | NM_000988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007930.hg.1 | NM_000988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009177.hg.1 | ENST00000427253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013493.hg.1 | ENST00000425821.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012286.hg.1 | ENST00000472241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009743.hg.1 | ENST00000433923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014041.hg.1 | ENST00000441321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006623.hg.1 | ENST00000253788.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006623.hg.1 | NM_000988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006623.hg.1 | TC17000544.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006827.hg.1 | ENST00000468034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157938.hg.1 | ENST00000253788.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157938.hg.1 | ENST00000587478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157938.hg.1 | ENST00000588830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157938.hg.1 | ENST00000589037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157938.hg.1 | ENST00000589913.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157938.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157938.hg.1 | NM_000988.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157938.hg.1 | ENST00000253788.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157938.hg.1 | ENST00000587478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157938.hg.1 | ENST00000588830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157938.hg.1 | ENST00000589037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157938.hg.1 | ENST00000589913.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157938.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157938.hg.1 | NM_000988.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157938.hg.1 | TC1700007930.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100013493.hg.1 | ENST00000425821.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000253788.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000253788.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000253788.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000253788.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000586277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000586277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000586277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000586277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000587478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000587478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000587478.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000588830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000588830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000588830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000589037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000589037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000589037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000589037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000589037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000589913.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000589913.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000589913.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000589913.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000589913.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000590864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000590864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000590864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000590864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | NM_000988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | NM_000988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | NM_000988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007930.hg.1 | NM_000988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010557.hg.1 | ENST00000434035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007618.hg.1 | ENST00000412199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007069.hg.1 | ENST00000477507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009177.hg.1 | ENST00000427253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012286.hg.1 | ENST00000472241.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157944.hg.1 | ENST00000253788.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157944.hg.1 | ENST00000586277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157944.hg.1 | ENST00000589037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157944.hg.1 | ENST00000589913.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157944.hg.1 | ENST00000590864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157944.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157944.hg.1 | NM_000988.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157944.hg.1 | ENST00000253788.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157944.hg.1 | ENST00000586277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157944.hg.1 | ENST00000589037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157944.hg.1 | ENST00000589913.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157944.hg.1 | ENST00000590864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157944.hg.1 | ENST00000593262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157944.hg.1 | NM_000988.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157944.hg.1 | TC1700007930.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500010536.hg.1 | ENST00000513087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006827.hg.1 | ENST00000468034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007781.hg.1 | ENST00000411843.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014041.hg.1 | ENST00000441321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009743.hg.1 | ENST00000433923.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 162467 | - | - |

RPL27
Genomic Map