| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000316649.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000316649.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000316649.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000316649.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000316649.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000316649.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000316649.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000316649.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000316649.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000316649.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000316649.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000316649.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367746.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367746.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367746.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367746.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367746.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367746.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367746.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367746.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367748.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367748.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367748.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367748.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367748.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367748.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367748.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367748.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367748.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367748.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367748.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367748.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000367748.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000460306.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000460306.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000460306.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000460306.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000461799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000461799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000461799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000468393.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000468393.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000468393.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000468393.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000468393.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000468393.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000468393.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000468393.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000541547.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000541547.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000541547.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000541547.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000541547.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000541547.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000541547.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000541547.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000541547.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000541547.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000541547.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
ENST00000541547.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278723.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278723.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278723.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278723.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278723.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278723.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278723.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278723.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278723.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278723.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278723.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278723.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278723.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278724.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278724.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278724.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278724.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278724.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278724.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278724.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278724.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278724.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278724.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278724.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_001278724.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_014432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_014432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_014432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_014432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_014432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_014432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_014432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_014432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_014432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_014432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_014432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_014432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_014432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013295.hg.1 |
NM_014432.3 |
Refseq |