Assay Details
| Gene Symbol: UQCRBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002308.1 | 1 | 575 | 152 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1420 | LINC01420 | Anti-sense | Chr. X - 56729285 - 56817571 on NCBI Build GRCh38 | ||
| Downstream | spindlin family member 3 | SPIN3 |
SPIN-3, TDRD27, bA445O16.1 |
238.8kb | Sense | Chr. X - 56976370 - 56995801 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001084.hg.1
Set Membership: |
|

UQCRBP1
Genomic Map