Assay Details
| Gene Symbol: GMPPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013334.3 | NP_037466.2 | 3 - 4 | 503 | 105 | |
| NM_021971.2 | NP_068806.1 | 3 - 4 | 503 | 105 | ||
| GenBank mRNA | AF135421.1 | - | 3 - 4 | 395 | 105 | |
| AK024319.1 | - | 3 - 4 | 480 | 105 | ||
| AK291700.1 | - | 3 - 4 | 478 | 105 | ||
| BC001141.1 | - | 3 - 4 | 442 | 3503869 | 105 | |
| BC008033.1 | - | 3 - 4 | 442 | 3355099 | 105 | |
| DA411701.1 | - | 3 - 4 | 503 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000481959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000481959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000481959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000481959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000481959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000481959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000481959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000481959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000495627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000495627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000495627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000495627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000495627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | ENST00000495627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207651.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207651.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207651.hg.1 | ENST00000481959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207651.hg.1 | ENST00000495627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207651.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207651.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207651.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207651.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207651.hg.1 | ENST00000481959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207651.hg.1 | ENST00000495627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207651.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207651.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207651.hg.1 | TC0300013980.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308375.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000308388.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000481959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000481959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000481959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000481959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000481959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000481959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000481959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000481959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000495627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000495627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000495627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000495627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000495627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | ENST00000495627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_013334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_021971.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013980.hg.1 | NM_021971.2 | Refseq |

GMPPB
Genomic Map