Assay Details
| Gene Symbol: PCMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252049.1 | NP_001238978.1 | 5 - 6 | 625 | 68 | |
| NM_001252050.1 | NP_001238979.1 | 4 - 5 | 520 | 68 | ||
| NM_001252051.1 | NP_001238980.1 | 4 - 5 | 520 | 68 | ||
| NM_001252052.1 | NP_001238981.1 | 4 - 5 | 520 | 68 | ||
| NM_001252053.1 | NP_001238982.1 | 5 - 6 | 625 | 68 | ||
| NM_005389.2 | NP_005380.2 | 5 - 6 | 625 | 68 | ||
| XM_011535868.2 | XP_011534170.1 | 5 - 6 | 826 | 68 | ||
| GenBank mRNA | AA442100.1 | - | 3 - 4 | 242 | 68 | |
| AK098739.1 | - | 4 - 5 | 503 | 68 | ||
| AK129778.1 | - | 5 - 6 | 541 | 68 | ||
| AK289724.1 | - | 5 - 6 | 593 | 68 | ||
| AK304532.1 | - | 4 - 5 | 441 | 68 | ||
| AL539512.3 | - | 4 - 5 | 492 | 68 | ||
| BC007501.2 | - | 5 - 6 | 567 | 3010742 | 68 | |
| BC008748.2 | - | 5 - 6 | 503 | 3346722 | 68 | |
| BC018569.2 | - | 1 - 2 | 110 | 68 | ||
| BE262147.1 | - | 4 - 5 | 405 | 68 | ||
| CX786396.1 | - | 5 - 6 | 511 | 68 | ||
| D13892.1 | - | 5 - 6 | 568 | 68 | ||
| D25545.1 | - | 5 - 6 | 491 | 68 | ||
| D25546.1 | - | 5 - 6 | 491 | 68 | ||
| D25547.1 | - | 5 - 6 | 491 | 68 | ||
| DC356391.1 | - | 5 - 6 | 608 | 68 | ||
| M93008.1 | - | 5 - 6 | 455 | 68 | ||
| M93009.1 | - | 1 - 2 | 61 | 68 | ||
More Information
Set Membership: |
|

PCMT1
Genomic Map