Assay Details
Gene Symbol: IFI27L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032036.2 | NP_114425.1 | 1 - 2 | 70 | 100 | |
GenBank mRNA | AF208232.1 | - | 1 - 2 | 36 | 100 | |
BC032626.1 | - | 1 - 2 | 29 | 5229498 | 100 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14001463.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF208232.1,BC032626.1,NM_032036.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR14016471.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14016471.hg.1 | ENST00000556727.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14016471.hg.1 | NM_032036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14016471.hg.1 | TC14001463.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR14016473.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14016473.hg.1 | NM_032036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14016473.hg.1 | TC14001463.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000553601.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000553601.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000553601.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000553601.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000554909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000554909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000554909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000554909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000554909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000555558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000555558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000555558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000555558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000555558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000555558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000556552.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000556552.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000556552.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000556552.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000556552.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000556552.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000556727.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000556727.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000556727.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000556727.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000556727.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | ENST00000556727.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | NM_032036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | NM_032036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | NM_032036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | NM_032036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | NM_032036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010041.hg.1 | NM_032036.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400174331.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400174331.hg.1 | NM_032036.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400174331.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400174331.hg.1 | NM_032036.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400174331.hg.1 | TC1400010041.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400174324.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400174324.hg.1 | ENST00000553601.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400174324.hg.1 | ENST00000555558.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400174324.hg.1 | ENST00000556552.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400174324.hg.1 | ENST00000556727.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400174324.hg.1 | NM_032036.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400174324.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400174324.hg.1 | ENST00000553601.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400174324.hg.1 | ENST00000555558.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400174324.hg.1 | ENST00000556552.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400174324.hg.1 | ENST00000556727.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400174324.hg.1 | NM_032036.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400174324.hg.1 | TC1400010041.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000238609.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000553601.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000553601.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000553601.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000553601.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000554909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000554909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000554909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000554909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000554909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000555558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000555558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000555558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000555558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000555558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000555558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000556552.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000556552.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000556552.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000556552.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000556552.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000556552.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000556727.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000556727.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000556727.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000556727.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000556727.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | ENST00000556727.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | NM_032036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | NM_032036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | NM_032036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | NM_032036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | NM_032036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010041.hg.1 | NM_032036.2 | Refseq |
