Assay Details
| Gene Symbol: PDE3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000921.4 | NP_000912.3 | 8 - 9 | 2046 | 74 | |
| NM_001244683.1 | NP_001231612.1 | 7 - 8 | 1303 | 74 | ||
| XM_017019420.1 | XP_016874909.1 | 8 - 9 | 2027 | 74 | ||
| XM_017019421.1 | XP_016874910.1 | 8 - 9 | 4744 | 74 | ||
| GenBank mRNA | AJ005036.1 | - | 8 - 9 | 2028 | 74 | |
| AK308317.1 | - | 7 - 8 | 1303 | 74 | ||
| BC117369.1 | - | 8 - 9 | 2046 | 40125920 | 74 | |
| BC117371.1 | - | 8 - 9 | 2046 | 40125922 | 74 | |
| M91667.2 | - | 8 - 9 | 2035 | 74 | ||
| U36798.1 | - | 8 - 9 | 2006 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000227.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
oocyte maturation
cAMP catabolic process
lipid metabolic process
diterpenoid metabolic process
cAMP-mediated signaling
cGMP-mediated signaling
regulation of meiotic nuclear division
response to drug
negative regulation of apoptotic process
negative regulation of vascular permeability
positive regulation of vascular permeability
response to cAMP
positive regulation of oocyte development
cellular response to cGMP
cellular response to transforming growth factor beta stimulus
3',5'-cyclic-AMP phosphodiesterase activity
cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
cAMP binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity
oocyte maturation
cAMP catabolic process
lipid metabolic process
diterpenoid metabolic process
cAMP-mediated signaling
cGMP-mediated signaling
regulation of meiotic nuclear division
response to drug
negative regulation of apoptotic process
negative regulation of vascular permeability
positive regulation of vascular permeability
response to cAMP
positive regulation of oocyte development
cellular response to cGMP
cellular response to transforming growth factor beta stimulus
3',5'-cyclic-AMP phosphodiesterase activity
cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
cAMP binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000542675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000542675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000542675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002762.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002762.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002762.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002762.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002762.hg.1 | TC12000227.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200149229.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149229.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149229.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149229.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149229.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149229.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149229.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149229.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149229.hg.1 | TC1200007037.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000359062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000542675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000542675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000542675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | ENST00000544307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_000921.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007037.hg.1 | NM_001244683.1 | Refseq |

PDE3A
Genomic Map