Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008050.hg.1 |
ENST00000312134.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008050.hg.1 |
ENST00000312134.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008050.hg.1 |
ENST00000312134.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008050.hg.1 |
ENST00000312134.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008050.hg.1 |
ENST00000312134.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008050.hg.1 |
ENST00000312134.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008050.hg.1 |
ENST00000312134.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008050.hg.1 |
NM_001323.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008050.hg.1 |
NM_001323.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008050.hg.1 |
NM_001323.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008050.hg.1 |
NM_001323.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008050.hg.1 |
NM_001323.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008050.hg.1 |
NM_001323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008050.hg.1 |
ENST00000312134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008050.hg.1 |
ENST00000312134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008050.hg.1 |
ENST00000312134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008050.hg.1 |
ENST00000312134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008050.hg.1 |
ENST00000312134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008050.hg.1 |
ENST00000312134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008050.hg.1 |
ENST00000312134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008050.hg.1 |
NM_001323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008050.hg.1 |
NM_001323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008050.hg.1 |
NM_001323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008050.hg.1 |
NM_001323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008050.hg.1 |
NM_001323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008050.hg.1 |
NM_001323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100158683.hg.1 |
ENST00000312134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100158683.hg.1 |
NM_001323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100158683.hg.1 |
ENST00000312134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100158683.hg.1 |
NM_001323.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100158683.hg.1 |
TC1100008050.hg.1 |
Clariom D TC |