Assay Details
| Gene Symbol: EEF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001961.3 | NP_001952.1 | 9 - 10 | 1430 | 63 | |
| GenBank mRNA | AK299225.1 | - | 9 - 10 | 1426 | 63 | |
| AY942181.1 | - | 8 - 9 | 1347 | 63 | ||
| BC006547.2 | - | 9 - 10 | 1415 | 63 | ||
| BC024689.1 | - | 2 - 3 | 325 | 63 | ||
| BC126259.1 | - | 9 - 10 | 1399 | 8991975 | 63 | |
| BC136313.1 | - | 9 - 10 | 1399 | 9020300 | 63 | |
| X51466.1 | - | 8 - 9 | 1347 | 63 | ||
| Z11692.1 | - | 8 - 9 | 1347 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001054.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
hematopoietic progenitor cell differentiation
response to ischemia
skeletal muscle contraction
translational elongation
aging
glial cell proliferation
response to fluoxetine
peptidyl-diphthamide biosynthetic process from peptidyl-histidine
response to estradiol
response to endoplasmic reticulum stress
skeletal muscle cell differentiation
response to hydrogen peroxide
response to ethanol
positive regulation of translation
response to folic acid
cell-cell adhesion
cellular response to brain-derived neurotrophic factor stimulus
positive regulation of cytoplasmic translation
p53 binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
5S rRNA binding
protein kinase binding
ribosome binding
poly(A) RNA binding
actin filament binding
cadherin binding involved in cell-cell adhesion
hematopoietic progenitor cell differentiation
response to ischemia
skeletal muscle contraction
translational elongation
aging
glial cell proliferation
response to fluoxetine
peptidyl-diphthamide biosynthetic process from peptidyl-histidine
response to estradiol
response to endoplasmic reticulum stress
skeletal muscle cell differentiation
response to hydrogen peroxide
response to ethanol
positive regulation of translation
response to folic acid
cell-cell adhesion
cellular response to brain-derived neurotrophic factor stimulus
positive regulation of cytoplasmic translation
p53 binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
5S rRNA binding
protein kinase binding
ribosome binding
poly(A) RNA binding
actin filament binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000384048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000384048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000594885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000594885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000594885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000594885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000596417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000596417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000596417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000600720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000600720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000600720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000600794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000600794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000600794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000600794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NR_002602.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NR_002602.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014411.hg.1 | ENST00000309311.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014411.hg.1 | ENST00000543343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014411.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014411.hg.1 | TC19001054.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900174387.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174387.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174387.hg.1 | TC1900009316.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900174387.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174387.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000384048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000384048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000594885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000594885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000594885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000594885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000596417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000596417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000596417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000600720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000600720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000600720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000600794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000600794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000600794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000600794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NR_002602.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NR_002602.1 | Refseq |

EEF2
Genomic Map