Assay Details
Gene Symbol: ETV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300974.1 | NP_001287903.1 | 5 - 6 | 1178 | 78 | |
NM_001304549.1 | NP_001291478.1 | 4 - 5 | 714 | 78 | ||
NM_014209.3 | NP_055024.2 | 6 - 7 | 1275 | 78 | ||
XM_005258652.2 | XP_005258709.1 | 5 - 6 | 1339 | 78 | ||
XM_011526624.2 | XP_011524926.1 | 5 - 6 | 1756 | 78 | ||
XM_017026472.1 | XP_016881961.1 | 4 - 5 | 1258 | 78 | ||
GenBank mRNA | AA885210.1 | - | 1 - 2 | 223 | 78 | |
AF000671.1 | - | 2 - 3 | 224 | 78 | ||
AK097439.1 | - | 5 - 6 | 1339 | 78 | ||
BC040908.1 | - | 5 - 6 | 1122 | 78 | ||
BC107123.1 | - | 7 - 8 | 721 | 78 | ||
BC107124.1 | - | 4 - 5 | 312 | 78 | ||
BC140746.1 | - | 6 - 7 | 965 | 9021614 | 78 | |
BC144448.1 | - | 6 - 7 | 963 | 9052974 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
