Assay Details
| Gene Symbol: IGDCC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020962.2 | NP_066013.1 | 3 - 4 | 776 | 91 | |
| XM_011521845.2 | XP_011520147.1 | 3 - 4 | 956 | 91 | ||
| XM_011521846.2 | XP_011520148.1 | 3 - 4 | 580 | 91 | ||
| XM_011521847.1 | XP_011520149.1 | 3 - 4 | 640 | 91 | ||
| XM_017022448.1 | XP_016877937.1 | 3 - 4 | 958 | 91 | ||
| XM_017022449.1 | XP_016877938.1 | 3 - 4 | 964 | 91 | ||
| GenBank mRNA | AB052622.1 | - | 3 - 4 | 776 | 91 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001566.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011521847.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000558819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000558819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000561309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000561309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000561309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014979.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014979.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014979.hg.1 | TC15001566.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000558819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000558819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000561309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000561309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000561309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172432.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172432.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172432.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172432.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172432.hg.1 | TC1500009796.hg.1 | Clariom D TC |

IGDCC4
Genomic Map