Assay Details
| Gene Symbol: IGDCC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020962.2 | NP_066013.1 | 10 - 11 | 2090 | 66 | |
| XM_011521845.2 | XP_011520147.1 | 10 - 11 | 2270 | 66 | ||
| XM_011521846.2 | XP_011520148.1 | 10 - 11 | 1894 | 66 | ||
| XM_011521847.1 | XP_011520149.1 | 10 - 11 | 1954 | 66 | ||
| XM_017022448.1 | XP_016877937.1 | 10 - 11 | 2272 | 66 | ||
| GenBank mRNA | AB046848.1 | - | 4 - 5 | 1150 | 66 | |
| AB052622.1 | - | 10 - 11 | 2090 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001566.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000558819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000558819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000561309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000561309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | ENST00000561309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014968.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014968.hg.1 | ENST00000356152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014968.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014968.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014968.hg.1 | TC15001566.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500172422.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172422.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172422.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172422.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172422.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172422.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172422.hg.1 | TC1500009796.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000352385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000558048.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000558819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000558819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000559327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000560319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000561309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000561309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | ENST00000561309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009796.hg.1 | NM_020962.2 | Refseq |

IGDCC4
Genomic Map