Assay Details
Gene Symbol: CALML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033429.2 | NP_219501.2 | 2 - 3 | 898 | 83 | |
GenBank mRNA | AK296804.1 | - | 2 - 3 | 120 | 83 | |
DB204573.1 | - | 2 - 3 | 122 | 83 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15001591.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK296804.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000354678.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000354678.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000354678.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000354678.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000448060.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000448060.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000448060.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000448060.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000448060.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000448060.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000448060.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000448060.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000478113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000478113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000478113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000478113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000478113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000478113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000478113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
