Assay Details
| Gene Symbol: PPP1R16A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005272350.2 | XP_005272407.1 | 5 - 6 | 1533 | 62 | |
| XM_011517349.2 | XP_011515651.1 | 4 - 5 | 384 | 62 | ||
| XM_017013912.1 | XP_016869401.1 | 6 - 7 | 1602 | 62 | ||
| XM_017013913.1 | XP_016869402.1 | 5 - 6 | 1566 | 62 | ||
| XM_017013914.1 | XP_016869403.1 | 6 - 7 | 1640 | 62 | ||
| XM_017013915.1 | XP_016869404.1 | 5 - 6 | 1597 | 62 | ||
| GenBank mRNA | AK090471.1 | - | 5 - 6 | 2663 | 62 | |
| BC007854.2 | - | 3 - 4 | 607 | 4298333 | 62 | |
| BC049841.1 | - | 3 - 4 | 859 | 4991947 | 62 | |
| BC053506.1 | - | 3 - 4 | 948 | 2823232 | 62 | |
| BT006845.1 | - | 3 - 4 | 479 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186508.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186508.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186508.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186508.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186508.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186508.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186508.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186508.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186508.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186508.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186508.hg.1 | TC0800012378.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800186509.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186509.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186509.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186509.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186509.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186509.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186509.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186509.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186509.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186509.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186509.hg.1 | TC0800012378.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |

PPP1R16A
Genomic Map