Assay Details
Gene Symbol: PPP1R16A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005272350.2 | XP_005272407.1 | 7 - 8 | 1758 | 116 | |
XM_011517349.2 | XP_011515651.1 | 6 - 7 | 609 | 116 | ||
XM_017013912.1 | XP_016869401.1 | 8 - 9 | 1827 | 116 | ||
XM_017013913.1 | XP_016869402.1 | 7 - 8 | 1791 | 116 | ||
XM_017013914.1 | XP_016869403.1 | 8 - 9 | 1865 | 116 | ||
XM_017013915.1 | XP_016869404.1 | 7 - 8 | 1822 | 116 | ||
GenBank mRNA | BC007854.2 | - | 5 - 6 | 832 | 4298333 | 116 |
BC049841.1 | - | 5 - 6 | 1084 | 4991947 | 116 | |
BC053506.1 | - | 5 - 6 | 1173 | 2823232 | 116 | |
BT006845.1 | - | 5 - 6 | 704 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800186515.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186515.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186515.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186515.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800186515.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186515.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186515.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186515.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800186515.hg.1 | TC0800012378.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000528430.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000529009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | ENST00000533829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012378.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800186513.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186513.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186513.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186513.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186513.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800186513.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186513.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186513.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186513.hg.1 | ENST00000532806.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186513.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800186513.hg.1 | TC0800012378.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800186514.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186514.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186514.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186514.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800186514.hg.1 | ENST00000292539.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186514.hg.1 | ENST00000435887.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186514.hg.1 | ENST00000526564.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800186514.hg.1 | NM_032902.6 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800186514.hg.1 | TC0800012378.hg.1 | Clariom D TC |
