Assay Details
Gene Symbol: CDKL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003948.3 | NP_003939.1 | 10 - 11 | 1945 | 83 | |
XM_017008810.1 | XP_016864299.1 | 10 - 11 | 1664 | 83 | ||
GenBank mRNA | AK312490.1 | - | 10 - 11 | 1936 | 83 | |
BC093646.1 | - | 10 - 11 | 1469 | 7939491 | 83 | |
BC093981.1 | - | 10 - 11 | 1469 | 7939826 | 83 | |
U35146.1 | - | 10 - 11 | 1795 | 83 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001299.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000506234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000506234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000506234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000506234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000506234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000515793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000515793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | ENST00000515793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04017674.hg.1 | TC04001299.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04017674.hg.1 | ENST00000307465.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04017674.hg.1 | ENST00000429927.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04017674.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000506234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000506234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000506234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000506234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000506234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000515793.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000515793.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | ENST00000515793.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011043.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400175071.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175071.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175071.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400175071.hg.1 | ENST00000307465.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175071.hg.1 | ENST00000429927.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175071.hg.1 | NM_003948.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400175071.hg.1 | TC0400011043.hg.1 | Clariom D TC |
