Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000506234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000506234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000506234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000506234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000506234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000515793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000515793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
ENST00000515793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017678.hg.1 |
TC04001299.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017678.hg.1 |
ENST00000307465.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017678.hg.1 |
ENST00000429927.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017678.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400175075.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400175075.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400175075.hg.1 |
ENST00000515793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400175075.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400175075.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400175075.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400175075.hg.1 |
ENST00000515793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400175075.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400175075.hg.1 |
TC0400011043.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000307465.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000429927.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000506234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000506234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000506234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000506234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000506234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000515793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000515793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
ENST00000515793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011043.hg.1 |
NM_003948.4 |
Refseq |